Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Ankrd40
- Entrez Gene ID
-
71452
- Gene Name
- ankyrin repeat domain 40
- Gene Aliases
-
1110011C06Rik, 5530600A18Rik, Gcap15
- Chromosome Location
-
Chr.11: 94328001 - 94341847 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
BC023133.1 |
- |
4 - 5 |
1051 |
95 |
View Full Product Details