The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_031869.2 | NP_114075.1 | 1 - 2 | 337 | 73 |
GenBank mRNA | AK153552.1 | - | 1 - 2 | 357 | 73 |
BC016398.1 | - | 1 - 2 | 337 | 73 |