The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_033314.3 | NP_201571.2 | 9 - 10 | 1423 | 56 |
GenBank mRNA | AF323958.1 | - | 9 - 10 | 1292 | 56 |
AK166724.1 | - | 9 - 10 | 1424 | 56 | |
AK169706.1 | - | 9 - 10 | 1426 | 56 |