The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001163529.1 | NP_001157001.1 | 2 - 3 | 477 | 82 |
NM_033615.2 | NP_291093.2 | 2 - 3 | 477 | 82 | |
GenBank mRNA | AB059632.1 | - | 2 - 3 | 477 | 82 |
AB059633.1 | - | 2 - 3 | 477 | 82 | |
AF472524.1 | - | 2 - 3 | 209 | 82 | |
AK053825.1 | - | 3 - 4 | 281 | 82 | |
AK086734.1 | - | 2 - 3 | 178 | 82 | |
AY054984.1 | - | 2 - 3 | 177 | 82 | |
AY382193.1 | - | 2 - 3 | 208 | 82 | |
AY382194.1 | - | 2 - 3 | 208 | 82 | |
BC138139.1 | - | 2 - 3 | 200 | 82 |