Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Adam33
Entrez Gene ID
110751
Gene Name
a disintegrin and metallopeptidase domain 33
Gene Aliases
Adaml
Chromosome Location
Chr.2: 131050817 - 131063814 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001163529.1 NP_001157001.1 18 - 19 2455 94
NM_033615.2 NP_291093.2 19 - 20 2533 94
GenBank mRNA AB059632.1 - 19 - 20 2533 94
AB059633.1 - 18 - 19 2455 94
AF386072.1 - 15 - 16 1897 94
AF472524.1 - 19 - 20 2265 94
AK048367.1 - 14 - 15 1889 94
AK053825.1 - 15 - 16 2952 94
BC138139.1 - 18 - 19 2178 94

View Full Product Details