The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_053169.2 | NP_444399.2 | 1 - 2 | 748 | 56 |
GenBank mRNA | AF449496.1 | - | 1 - 2 | 499 | 56 |
AK053139.1 | - | 1 - 2 | 562 | 56 | |
BC052821.1 | - | 2 - 3 | 450 | 56 |