The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_053253.3 | NP_444483.2 | 2 - 3 | 350 | 76 |
GenBank mRNA | AK132332.1 | - | 2 - 3 | 359 | 76 |
BC052357.1 | - | 2 - 3 | 350 | 76 |