Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Zmynd10
Entrez Gene ID
114602
Gene Name
zinc finger, MYND domain containing 10
Gene Aliases
Blu
Chromosome Location
Chr.9: 107545991 - 107551321 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_053253.3 NP_444483.2 7 - 8 847 83
GenBank mRNA AF123386.1 - 5 - 6 478 83
AK132332.1 - 7 - 8 856 83
BC052357.1 - 7 - 8 847 83

View Full Product Details