The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_133345.2 | NP_579923.1 | 7 - 8 | 714 | 111 |
GenBank mRNA | AK004347.1 | - | - | 714 | 111 |
AK009267.1 | - | 7 - 8 | 715 | 111 | |
AK031633.1 | - | 7 - 8 | 722 | 111 | |
AK050522.1 | - | 7 - 8 | 727 | 111 | |
AK184719.1 | - | 1 - 2 | 61 | 111 | |
AK186414.1 | - | 1 - 2 | 61 | 111 | |
AY035880.1 | - | 6 - 7 | 906 | 111 | |
AY035881.1 | - | 6 - 7 | 936 | 111 | |
AY036107.1 | - | 7 - 8 | 583 | 111 | |
BC009127.1 | - | 7 - 8 | 702 | 111 | |
BC082781.1 | - | 2 - 3 | 3134 | 111 |