The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_134151.4 | NP_598912.4 | 1 - 2 | 277 | 82 |
GenBank mRNA | AK043837.1 | - | 1 - 2 | 201 | 82 |
AK076634.1 | - | 1 - 2 | 122 | 82 | |
AK145356.1 | - | 1 - 2 | 170 | 82 | |
AK169708.1 | - | 1 - 2 | 53 | 82 | |
BC013552.1 | - | 1 - 2 | 82 | ||
BC022143.1 | - | 1 - 2 | 82 | ||
BC026615.1 | - | 1 - 2 | 82 | ||
BY287648.1 | - | 1 - 2 | 277 | 82 | |
BY295788.1 | - | 1 - 2 | 277 | 82 |