The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_134151.4 | NP_598912.4 | 12 - 13 | 1703 | 69 |
GenBank mRNA | AK028785.1 | - | 12 - 13 | 1565 | 69 |
AK043837.1 | - | 12 - 13 | 1627 | 69 | |
AK076634.1 | - | 12 - 13 | 1548 | 69 | |
AK139579.1 | - | 12 - 13 | 1548 | 69 | |
AK145356.1 | - | 12 - 13 | 1596 | 69 | |
AK151880.1 | - | 12 - 13 | 1555 | 69 | |
AK169708.1 | - | 12 - 13 | 1479 | 69 | |
BC013552.1 | - | 12 - 13 | 1634 | 69 | |
BC022143.1 | - | 12 - 13 | 1598 | 69 | |
BC026615.1 | - | 12 - 13 | 1610 | 69 |