The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001159536.1 | NP_001153008.1 | 11 - 12 | 2462 | 64 |
NM_001159537.1 | NP_001153009.1 | 11 - 12 | 2521 | 64 | |
NM_138305.3 | NP_612178.2 | 10 - 11 | 2139 | 64 | |
GenBank mRNA | AF458089.1 | - | 10 - 11 | 2111 | 64 |
AK122298.1 | - | 11 - 12 | 2433 | 64 | |
BC057113.1 | - | 11 - 12 | 2450 | 64 | |
BC057316.1 | - | 11 - 12 | 2351 | 64 |