The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_144929.2 | NP_659178.1 | 2 - 3 | 957 | 85 |
GenBank mRNA | AK080715.1 | - | 2 - 3 | 273 | 85 |
AY428631.1 | - | 1 - 2 | 169 | 85 | |
AY527275.1 | - | 2 - 3 | 957 | 85 | |
AY527276.1 | - | 3 - 4 | 412 | 85 | |
BC022744.1 | - | 1 - 2 | 529 | 85 | |
BC023051.1 | - | 1 - 2 | 485 | 85 | |
BC027079.1 | - | 1 - 2 | 485 | 85 |