The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001290299.1 | NP_001277228.1 | 9 - 10 | 1346 | 62 |
NM_144935.1 | NP_659184.1 | 8 - 9 | 1302 | 62 | |
GenBank mRNA | AK082453.1 | - | 9 - 10 | 1309 | 62 |
AY438023.1 | - | 8 - 9 | 1302 | 62 | |
BC018242.1 | - | 8 - 9 | 1302 | 62 | |
BC023082.1 | - | 9 - 10 | 1323 | 62 |