The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001174155.1 | NP_001167626.1 | 8 - 9 | 1097 | 76 |
NM_145149.4 | NP_660131.2 | 8 - 9 | 1097 | 76 | |
NR_045676.1 | 8 - 9 | 1097 | 76 | ||
GenBank mRNA | AF331457.1 | - | 8 - 9 | 1097 | 76 |
AK049562.1 | - | 7 - 8 | 840 | 76 | |
AK087348.1 | - | 8 - 9 | 1050 | 77 | |
AK089282.1 | - | 8 - 9 | 1044 | 76 | |
AK154746.1 | - | 8 - 9 | 1087 | 76 | |
AK162667.1 | - | 8 - 9 | 1098 | 76 | |
AK165106.1 | - | 7 - 8 | 835 | 76 | |
AY040628.1 | - | 8 - 9 | 1097 | 76 | |
AY196476.1 | - | 8 - 9 | 957 | 76 |