Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Oas2
Entrez Gene ID
246728
Gene Name
2'-5' oligoadenylate synthetase 2
Gene Aliases
Oasl11
Chromosome Location
Chr.5: 120730333 - 120749884 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AF418010.1 - 10 - 11 2175 95

View Full Product Details