The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_145470.3 | NP_663445.2 | 1 - 2 | 307 | 91 |
GenBank mRNA | AK145208.1 | - | 1 - 2 | 201 | 91 |
AK220300.1 | - | 1 - 2 | 141 | 91 | |
BC004774.1 | - | 1 - 2 | 312 | 91 | |
DV649395.1 | - | 1 - 2 | 274 | 91 |