The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_145592.2 | NP_663567.1 | 3 - 4 | 558 | 73 |
GenBank mRNA | BC018400.1 | - | 3 - 4 | 558 | 73 |