Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Klhdc4
Entrez Gene ID
234825
Gene Name
kelch domain containing 4
Gene Aliases
AA408426, AV352552, BC012312, G430025P05Rik
Chromosome Location
Chr.8: 121796308 - 121829577 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_145605.2 NP_663580.2 2 - 3 279 86
GenBank mRNA AK089954.1 - 2 - 3 242 86
AK161674.1 - 2 - 3 255 86
BC012312.1 - 2 - 3 279 86
BC023738.1 - 2 - 3 250 86
BC058359.1 - 2 - 3 250 86
BY086869.1 - 2 - 3 265 86

View Full Product Details