The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_145937.3 | NP_666049.2 | 3 - 4 | 543 | 60 |
GenBank mRNA | AK151874.1 | - | 3 - 4 | 544 | 60 |
AK159192.1 | - | 3 - 4 | 532 | 60 | |
AK160917.1 | - | 3 - 4 | 525 | 60 | |
AK161203.1 | - | 3 - 4 | 536 | 60 | |
AK194677.1 | - | 2 - 3 | 122 | 60 | |
AK198586.1 | - | 2 - 3 | 122 | 60 | |
BC026981.1 | - | 3 - 4 | 516 | 60 |