The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_145962.2 | NP_666074.1 | 5 - 6 | 1362 | 100 |
GenBank mRNA | AK039865.1 | - | 5 - 6 | 1364 | 100 |
AK136305.1 | - | 5 - 6 | 1052 | 100 | |
AK163377.1 | - | 4 - 5 | 749 | 100 | |
BC027089.1 | - | 5 - 6 | 974 | 100 | |
BC032188.1 | - | 5 - 6 | 1408 | 100 |