The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_146025.2 | NP_666137.1 | 9 - 10 | 1805 | 86 |
GenBank mRNA | AK044765.1 | - | 9 - 10 | 1334 | 86 |
AK047184.1 | - | 9 - 10 | 1782 | 86 | |
AK049753.1 | - | 9 - 10 | 1808 | 86 | |
BC034054.1 | - | 9 - 10 | 1329 | 86 | |
BC049954.1 | - | 9 - 10 | 1362 | 86 |