Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
B4galt7
Entrez Gene ID
218271
Gene Name
xylosylprotein beta1,4-galactosyltransferase, polypeptide 7 (galactosyltransferase I)
Gene Aliases
-
Chromosome Location
Chr.13: 55599901 - 55610445 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_146045.1 NP_666157.1 4 - 5 756 84
GenBank mRNA AK141739.1 - 4 - 5 720 84
AK146855.1 - 4 - 5 803 84
AK171600.1 - 4 - 5 772 84
BC027195.1 - 4 - 5 756 84

View Full Product Details