The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_146248.2 | NP_666360.2 | 4 - 5 | 699 | 69 |
GenBank mRNA | BC031416.1 | - | 5 - 6 | 788 | 69 |
BM947393.1 | - | 4 - 5 | 611 | 69 |