The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001110513.1 | NP_001103983.1 | 15 - 16 | 1803 | 66 |
GenBank mRNA | AF387632.1 | - | 15 - 16 | 1700 | 66 |
AF387633.1 | - | 15 - 16 | 1803 | 66 | |
AF387634.1 | - | 15 - 16 | 1687 | 66 | |
AK149248.1 | - | 4 - 5 | 827 | 66 | |
BC138119.1 | - | 15 - 16 | 1803 | 66 | |
BC145155.1 | - | 16 - 17 | 1889 | 66 |