The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001038590.1 | NP_001033679.1 | 1 - 2 | 331 | 64 |
NM_153105.7 | NP_694745.1 | 1 - 2 | 359 | 64 | |
GenBank mRNA | AF249889.1 | - | 1 - 2 | 199 | 64 |
AF486651.2 | - | 1 - 2 | 331 | 64 | |
AK032743.1 | - | 1 - 2 | 292 | 64 | |
AK143945.1 | - | 1 - 2 | 360 | 64 | |
BC114989.1 | - | 1 - 2 | 292 | 64 | |
BC115827.1 | - | 1 - 2 | 292 | 64 |