The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_153118.3 | NP_694758.2 | 9 - 10 | 1152 | 67 |
GenBank mRNA | BC034510.1 | - | 4 - 5 | 600 | 67 |