Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Abca8a
Entrez Gene ID
217258
Gene Name
ATP-binding cassette, sub-family A (ABC1), member 8a
Gene Aliases
-
Chromosome Location
Chr.11: 110025634 - 110097583 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_153145.4 NP_694785.3 29 - 30 3903 55
GenBank mRNA AB255881.1 - 28 - 29 3822 55
AB255882.1 - 29 - 30 3910 55
AF498360.1 - 28 - 29 3831 55
AK031843.1 - 10 - 11 1134 55
AY732492.1 - 28 - 29 3831 55
BC026496.1 - 9 - 10 1028 55
BC060032.1 - 11 - 12 1397 55

View Full Product Details