The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_153154.2 | NP_694794.1 | 6 - 7 | 1264 | 84 |
GenBank mRNA | AF421891.1 | - | 6 - 7 | 1291 | 84 |
AK132112.1 | - | 6 - 7 | 1266 | 84 | |
BC152324.1 | - | 6 - 7 | 1213 | 84 | |
BC152325.1 | - | 6 - 7 | 1213 | 84 |