The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_153159.2 | NP_694799.1 | 5 - 6 | 1075 | 88 |
GenBank mRNA | AK142501.1 | - | 5 - 6 | 1067 | 88 |
AK149698.1 | - | 5 - 6 | 1074 | 88 | |
AK152196.1 | - | 5 - 6 | 1032 | 88 | |
AK152518.1 | - | 3 - 4 | 611 | 88 | |
AK154363.1 | - | 5 - 6 | 1048 | 88 | |
AK161150.1 | - | 5 - 6 | 1031 | 88 | |
AK161733.1 | - | 5 - 6 | 1030 | 88 | |
AK172357.1 | - | 5 - 6 | 1032 | 88 | |
AY920404.1 | - | 4 - 5 | 926 | 88 | |
BC006817.1 | - | 2 - 3 | 230 | 88 | |
BC036563.1 | - | 5 - 6 | 1034 | 88 |