The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_153167.2 | NP_694807.2 | 1 - 2 | 626 | 62 |
GenBank mRNA | AK045241.1 | - | 1 - 2 | 546 | 62 |
AK083428.1 | - | 1 - 2 | 227 | 62 | |
AK157345.1 | - | 1 - 2 | 561 | 62 | |
AK165246.1 | - | 1 - 2 | 626 | 62 | |
AK170534.1 | - | 1 - 2 | 631 | 62 | |
BC027317.1 | - | 1 - 2 | 586 | 62 | |
BC085101.1 | - | 1 - 2 | 483 | 62 |