The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_153167.2 | NP_694807.2 | 4 - 5 | 1140 | 92 |
GenBank mRNA | AK045241.1 | - | 5 - 6 | 1115 | 92 |
AK083428.1 | - | 4 - 5 | 741 | 92 | |
AK157345.1 | - | 4 - 5 | 1076 | 92 | |
AK170534.1 | - | 5 - 6 | 1200 | 92 | |
AK194080.1 | - | 1 - 2 | 158 | 92 | |
AK213618.1 | - | 1 - 2 | 158 | 92 | |
BC027317.1 | - | 4 - 5 | 1100 | 92 | |
BC085101.1 | - | 4 - 5 | 997 | 92 | |
CN533345.1 | - | 2 - 3 | 224 | 92 |