The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_153287.3 | NP_695019.1 | 2 - 3 | 272 | 77 |
GenBank mRNA | AB091687.1 | - | 1 - 2 | 244 | 77 |
AK029893.1 | - | 2 - 3 | 539 | 77 | |
AK171658.1 | - | 1 - 2 | 338 | 77 | |
BC050066.1 | - | 2 - 3 | 272 | 77 |