The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001164833.1 | NP_001158305.1 | 10 - 11 | 1153 | 107 |
NM_001164834.1 | NP_001158306.1 | 10 - 11 | 1153 | 107 | |
NM_153400.4 | NP_700449.2 | 10 - 11 | 1153 | 107 | |
GenBank mRNA | AB094663.1 | - | 10 - 11 | 1073 | 107 |
AB094664.1 | - | 10 - 11 | 1073 | 107 | |
AK141196.1 | - | 10 - 11 | 1112 | 107 | |
AK158334.1 | - | 10 - 11 | 1131 | 107 | |
AY044240.2 | - | 10 - 11 | 1329 | 107 | |
BC106964.1 | - | 10 - 11 | 960 | 107 |