The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001145826.1 | NP_001139298.1 | 11 - 12 | 2974 | 83 |
NM_153406.3 | NP_700455.3 | 8 - 9 | 2745 | 83 | |
GenBank mRNA | AB093236.1 | - | 11 - 12 | 2974 | 83 |
AK041258.1 | - | 1 - 2 | 989 | 83 | |
AY884297.1 | - | 8 - 9 | 2746 | 83 | |
BC037464.1 | - | 5 - 6 | 1832 | 83 | |
BC072595.1 | - | 7 - 8 | 2584 | 83 |