The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001165954.1 | NP_001159426.1 | 3 - 4 | 359 | 122 |
NM_001165955.1 | NP_001159427.1 | 3 - 4 | 374 | 122 | |
NM_001165956.1 | NP_001159428.1 | 3 - 4 | 359 | 122 | |
NM_153421.2 | NP_700470.2 | 3 - 4 | 374 | 122 | |
GenBank mRNA | AJ414610.1 | - | 3 - 4 | 360 | 122 |
AK037348.1 | - | 3 - 4 | 350 | 122 | |
AK087339.1 | - | 3 - 4 | 365 | 122 | |
AK166208.1 | - | 3 - 4 | 350 | 122 | |
AK166296.1 | - | 3 - 4 | 360 | 122 | |
BC132533.1 | - | 3 - 4 | 343 | 122 | |
BC145123.1 | - | 3 - 4 | 343 | 122 | |
BC145124.1 | - | 3 - 4 | 343 | 122 |