The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_153423.6 | NP_700472.1 | 6 - 7 | 803 | 92 |
GenBank mRNA | AK035717.1 | - | 1 - 2 | 111 | 92 |
AK085458.1 | - | 6 - 7 | 804 | 92 | |
AK150140.1 | - | 6 - 7 | 872 | 92 | |
AK172187.1 | - | 6 - 7 | 835 | 92 | |
AK211371.1 | - | 1 - 2 | 86 | 92 | |
AK218674.1 | - | 1 - 2 | 86 | 92 | |
AY135643.1 | - | 5 - 6 | 777 | 92 |