The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001165983.1 | NP_001159455.1 | 17 - 18 | 2110 | 107 |
NM_001165984.1 | NP_001159456.1 | 17 - 18 | 2070 | 107 | |
NM_001165985.1 | NP_001159457.1 | 17 - 18 | 2128 | 107 | |
NM_001165986.1 | NP_001159458.1 | 17 - 18 | 2103 | 107 | |
NM_001165987.1 | NP_001159459.1 | 16 - 17 | 2035 | 107 | |
NM_001165988.1 | NP_001159460.1 | 16 - 17 | 2010 | 107 | |
NM_028475.2 | NP_082751.1 | 17 - 18 | 2095 | 107 | |
NM_153489.2 | NP_705693.2 | 16 - 17 | 2035 | 107 | |
GenBank mRNA | AF276967.1 | - | 16 - 17 | 1970 | 107 |
AK030218.1 | - | 17 - 18 | 2036 | 107 | |
AK031934.1 | - | 16 - 17 | 2015 | 107 | |
AK048286.1 | - | 17 - 18 | 2111 | 107 | |
AK079895.1 | - | 17 - 18 | 2067 | 107 | |
AK086314.1 | - | 2 - 3 | 1496 | 107 | |
AK088702.1 | - | 17 - 18 | 2126 | 107 | |
AK122210.1 | - | 4 - 5 | 473 | 107 | |
AK162166.1 | - | 2 - 3 | 123 | 107 | |
AK213629.1 | - | 2 - 3 | 109 | 107 | |
BC023906.1 | - | 16 - 17 | 2026 | 107 | |
BC029075.1 | - | 17 - 18 | 2085 | 107 | |
BC050910.1 | - | 16 - 17 | 2017 | 107 |