Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Ankrd23
- Entrez Gene ID
-
78321
- Gene Name
- ankyrin repeat domain 23
- Gene Aliases
-
1110058D09Rik, 2310041C05Rik, AI853437, Darp, MARP3
- Chromosome Location
-
Chr.1: 36530188 - 36536883 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
BC022973.1 |
- |
- |
150 |
54 |
View Full Product Details