The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_153570.2 | NP_705798.2 | 2 - 3 | 279 | 89 |
GenBank mRNA | AK052601.1 | - | 2 - 3 | 279 | 89 |
AK088355.1 | - | 2 - 3 | 280 | 89 | |
AK090024.1 | - | 2 - 3 | 292 | 89 | |
AK146006.1 | - | 2 - 3 | 275 | 89 | |
AK172310.1 | - | 2 - 3 | 258 | 89 | |
BC024616.1 | - | 2 - 3 | 269 | 89 |