Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Noc4l
Entrez Gene ID
100608
Gene Name
nucleolar complex associated 4 homolog (S. cerevisiae)
Gene Aliases
AI326906
Chromosome Location
Chr.5: 110648419 - 110653382 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_153570.2 NP_705798.2 4 - 5 491 88
GenBank mRNA AK052601.1 - 4 - 5 491 88
AK088355.1 - 4 - 5 492 88
AK090024.1 - 4 - 5 504 88
AK146006.1 - 4 - 5 487 88
AK172310.1 - 4 - 5 470 88
BC024616.1 - 4 - 5 481 88

View Full Product Details