The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_153573.1 | NP_705801.1 | 1 - 2 | 279 | 83 |
GenBank mRNA | AK030620.1 | - | 1 - 2 | 329 | 83 |
AK081692.1 | - | 1 - 2 | 306 | 83 | |
BC024846.1 | - | - | 616 | 83 | |
BC029109.1 | - | 1 - 2 | 279 | 83 | |
BC034313.1 | - | 1 - 2 | 304 | 83 |