Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Gpm6a
Entrez Gene ID
234267
Gene Name
glycoprotein m6a
Gene Aliases
Gpm6, M6A
Chromosome Location
Chr.8: 54779434 - 55060878 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_153581.6 NP_705809.1 1 - 2 589 111
GenBank mRNA AK053538.1 - 1 - 2 143 111
AK081877.1 - 1 - 2 475 111
AK147245.1 - 1 - 2 376 111
AK147280.1 - 1 - 2 376 111
BC024759.1 - 1 - 2 116 111
BC029683.1 - 1 - 2 158 111
BC029711.1 - 1 - 2 112 111
BC033357.1 - 1 - 2 158 111
BC033394.1 - 1 - 2 116 111
BC043130.1 - 1 - 2 404 111
S65735.1 - 1 - 2 115 111

View Full Product Details