The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001122667.2 | NP_001116139.1 | 5 - 6 | 452 | 76 |
NM_153588.3 | NP_705816.2 | 3 - 4 | 466 | 76 | |
GenBank mRNA | AF532598.1 | - | 3 - 4 | 248 | 76 |
AK220377.1 | - | 3 - 4 | 248 | 76 | |
BC131644.1 | - | 3 - 4 | 248 | 76 | |
BC131645.1 | - | 3 - 4 | 246 | 76 |