Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Snx17
Entrez Gene ID
266781
Gene Name
sorting nexin 17
Gene Aliases
5830447M19Rik, AI790646, D5Ertd260e, b2b1625.1Clo, mKIAA0064
Chromosome Location
Chr.5: 31193287 - 31198900 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_153680.2 NP_710147.1 4 - 5 424 91
GenBank mRNA AK047265.1 - 4 - 5 424 91
AK077650.1 - 4 - 5 426 91
AK077780.1 - 4 - 5 426 91
AK156299.1 - 4 - 5 475 91
AK219724.1 - 1 - 2 69 91
BC023732.1 - 4 - 5 440 91
BC026571.1 - 4 - 5 424 91
BC029675.1 - 4 - 5 411 91

View Full Product Details