The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_153783.4 | NP_722478.2 | 1 - 2 | 201 | 91 |
GenBank mRNA | AF226656.2 | - | 1 - 2 | 220 | 91 |
AK030664.1 | - | 1 - 2 | 202 | 91 | |
AM049402.1 | - | 1 - 2 | 193 | 91 | |
BC082783.1 | - | 1 - 2 | 200 | 91 |