Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Bclaf1
Entrez Gene ID
72567
Gene Name
BCL2-associated transcription factor 1
Gene Aliases
2610102K23Rik, 2700025J07Rik, 2810454G14Rik, 5730534O06Rik, AI450190, AW556225, Btf, mKIAA0164
Chromosome Location
Chr.10: 20311461 - 20342503 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001025392.1 NP_001020563.1 11 - 12 2793 69
NM_153787.2 NP_722482.1 11 - 12 2787 69
GenBank mRNA AK079110.1 - 7 - 8 1043 69
AK129071.1 - 8 - 9 2077 69
AK149724.1 - 11 - 12 2746 69
AK150017.1 - 11 - 12 2746 69
BC029682.1 - 11 - 12 2725 69
BC032973.1 - 2 - 3 468 69
BC034300.1 - 11 - 12 2719 69
CN530237.1 - 2 - 3 178 69

View Full Product Details