The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_153801.3 | NP_722496.2 | 1 - 2 | 424 | 115 |
GenBank mRNA | AF548365.1 | - | 1 - 2 | 425 | 115 |
AK052276.1 | - | 1 - 2 | 263 | 115 | |
AK052284.1 | - | 1 - 2 | 427 | 115 | |
AK052302.1 | - | 1 - 2 | 129 | 115 | |
AK052378.1 | - | 1 - 2 | 187 | 115 | |
BC107366.1 | - | 1 - 2 | 337 | 115 | |
BC107367.1 | - | 1 - 2 | 337 | 115 |