Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Arid4a
Entrez Gene ID
238247
Gene Name
AT rich interactive domain 4A (RBP1-like)
Gene Aliases
A630009N03, A630067N03Rik, MmRBBP1, Rbbp1
Chromosome Location
Chr.12: 71015967 - 71099351 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001081195.1 NP_001074664.1 2 - 3 254 82
GenBank mRNA AK042186.1 - 1 - 2 443 82
AK050594.1 - 2 - 3 194 82
AY116674.1 - 2 - 3 141 82
BC025436.1 - 2 - 3 242 82
BC058209.1 - 2 - 3 171 82
BC080780.1 - 2 - 3 189 82
BC157975.1 - 1 - 2 56 82
BC158109.1 - 1 - 2 56 82

View Full Product Details