The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001291354.1 | NP_001278283.1 | 2 - 3 | 563 | 111 |
NM_007491.2 | NP_031517.2 | 2 - 3 | 615 | 111 | |
NR_111935.1 | 2 - 3 | 563 | 111 | ||
GenBank mRNA | AJ295843.1 | - | 1 - 2 | 66 | 111 |
AJ297547.1 | - | 1 - 2 | 66 | 111 | |
AJ297548.1 | - | 1 - 2 | 66 | 111 | |
AK132854.1 | - | 2 - 3 | 616 | 111 | |
BC116823.1 | - | 2 - 3 | 427 | 111 | |
BC116849.1 | - | 2 - 3 | 427 | 111 | |
BC144825.1 | - | 2 - 3 | 427 | 111 | |
BC144826.1 | - | 2 - 3 | 375 | 111 | |
BC144827.1 | - | 1 - 2 | 614 | 111 | |
BC144836.1 | - | 2 - 3 | 375 | 111 | |
U60881.1 | - | - | 448 | 111 |